4-Amino-2-nitrobenzaldehyde 95%

Supplier: Ambeed
Warning

A257370-100MG A257370-1G A257370-250MG
77140-654EA 147.28 USD
77140-654 77140-656 77140-658
4-Amino-2-nitrobenzaldehyde 95%
4-Amino-2-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₆N₂O₃
MDL Number: MFCD13178316
CAS Number: 102170-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR