(R,R)-N,N'-Bis(trifluoromethanesulfonyl)-1,2-diphenylethylenediamine 97%

Supplier: Ambeed
Warning

Synonyms: N,N'-((1R,2R)-1,2-Diphenylethane-1,2-diyl)bis(1,1,1-trifluoromethanesulphonamide), (R,R)-1,2-Bis(trifluoromethanesulphonamido)-1,2-diphenylethane

A257257-10G A257257-1G A257257-5G A257257-250MG
77140-648EA 123.07 USD
77140-648 77140-650 77140-652 77746-414
(R,R)-N,N'-Bis(trifluoromethanesulfonyl)-1,2-diphenylethylenediamine 97%
(R,R)-N,N'-Bis(trifluoromethanesulfonyl)-1,2-diphenylethylenediamine

Precaución: for Research Use Only

Formula: C₁₆H₁₄F₆N₂O₄S₂
MW: 476.41 g/mol
MDL Number: MFCD01321158
CAS Number: 121788-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR