(3-((6-(2-Methoxyphenyl)pyrimidin-4-yl)amino)phenyl)methanesulfonamide 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
LDC000067
A256876-10MG
A256876-50MG
A256876-5MG
A256876-1MG
77140-636EA
21.22
USD
77140-636
77140-638
77140-640
77616-076
(3-((6-(2-Methoxyphenyl)pyrimidin-4-yl)amino)phenyl)methanesulfonamide 98%
(3-((6-(2-Methoxyphenyl)pyrimidin-4-yl)amino)phenyl)methanesulfonamide
Precaución: for Research Use Only
Formula:
C₁₈H₁₈N₄O₃S MW: 370.43 g/mol |
MDL Number:
MFCD28137788 CAS Number: 1073485-20-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...