(1S,4R)-Boc-4-aminocyclopent-2-enecarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: (1S,4R)-4-((tert-Butoxycarbonyl)amino)cyclopent-2-enecarboxylic acid

A255245-100MG A255245-1G A255245-250MG A255245-25G A255245-5G
77140-612EA 14.42 USD
77140-612 77140-614 77140-616 77140-618 77140-620
(1S,4R)-Boc-4-aminocyclopent-2-enecarboxylic acid 97%
(1S,4R)-Boc-4-aminocyclopent-2-enecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₄
MW: 227.26 g/mol
MDL Number: MFCD00211287
CAS Number: 151907-79-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR