Balicatib 98+%

Supplier: Ambeed
Warning

A254787-100MG A254787-10MG A254787-1MG A254787-50MG A254787-5MG
77140-600EA 216.3 USD
77140-600 77140-602 77140-604 77140-606 77140-608
Balicatib 98+%
Balicatib

Precaución: for Research Use Only

Formula: C₂₃H₃₃N₅O₂
MDL Number: MFCD19443790
CAS Number: 354813-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR