2,3-Dibromo-5-nitropyridine 95%

Supplier: Ambeed
Warning

A254784-100G A254784-10G A254784-1G A254784-25G A254784-5G
77140-590EA 387.23 USD
77140-590 77140-592 77140-594 77140-596 77140-598
2,3-Dibromo-5-nitropyridine 95%
2,3-Dibromo-5-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₂Br₂N₂O₂
MW: 281.8896 g/mol
MDL Number: MFCD00233994
CAS Number: 15862-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR