2,6-Dimethoxyanthracene 95%

Supplier: Ambeed
Warning

A254691-100MG A254691-1G A254691-250MG A254691-5G
77140-582EA 55.62 USD
77140-582 77140-584 77140-586 77140-588
2,6-Dimethoxyanthracene 95%
2,6-Dimethoxyanthracene

Precaución: for Research Use Only

Formula: C₁₆H₁₄O₂
MDL Number: MFCD28969170
CAS Number: 36319-03-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR