3H-1,2-Benzodithiol-one-1,1-dioxide (beaucage reagent) 95%

Supplier: Ambeed
Warning

A254406-1G A254406-1MG A254406-250MG A254406-5G
77140-570EA 34 USD
77140-570 77140-572 77140-574 77685-724
3H-1,2-Benzodithiol-one-1,1-dioxide (beaucage reagent) 95%
3H-1,2-Benzodithiol-one 1,1-dioxide (Beaucage Reagent)

Precaución: for Research Use Only

Formula: C₇H₄O₃S₂
MDL Number: MFCD00132960
CAS Number: 66304-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR