3-Iodotoluene 98%

Supplier: Ambeed
Warning

A254232-1000G A254232-100G A254232-10G A254232-25G A254232-500G A254232-5G
77140-558EA 301.79 USD
77140-558 77140-560 77140-562 77140-564 77140-566 77140-568
3-Iodotoluene 98%
3-Iodotoluene

Precaución: for Research Use Only

Formula: C₇H₇I
MDL Number: MFCD00001050
CAS Number: 625-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR