(R)-(+)-1-Phenylethanol 98%

Supplier: Ambeed
Danger

Synonyms: (R)-1-Phenylethanol, (R)-(+)-α-Methylbenzyl alcohol, (R)-(+)-sec-Phenethyl alcohol

A253233-10G A253233-1G A253233-25G A253233-5G
77140-540EA 13.8 USD
77140-540 77140-542 77140-544 77140-546
(R)-(+)-1-Phenylethanol 98%
(R)-(+)-1-Phenylethanol

Precaución: for Research Use Only

Formula: C₈H₁₀O
MW: 122.17 g/mol
MDL Number: MFCD00064263
CAS Number: 1517-69-7
UN: 2922
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR