1-Oxo-2,3-dihydro-1H-indene-4-carbonitrile 97%
Supplier: Ambeed
|
Warning
|
A253109-1G
A253109-250MG
A253109-25G
A253109-5G
77140-532EA
27.59
USD
77140-532
77140-534
77140-536
77140-538
1-Oxo-2,3-dihydro-1H-indene-4-carbonitrile 97%
1-Oxo-2,3-dihydro-1H-indene-4-carbonitrile
Precaución: for Research Use Only
Formula:
C₁₀H₇NO MW: 157.1700 g/mol |
MDL Number:
MFCD01111989 CAS Number: 60899-34-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...