2-Chlorobenzophenone 98%

Supplier: Ambeed
Warning

Synonyms: o-Chlorobenzophenone, (2-Chlorophenyl)(phenyl)methanone

A252600-1000G A252600-100G A252600-10G A252600-25G A252600-500G
77140-522EA 99.73 USD
77140-522 77140-524 77140-526 77140-528 77140-530
2-Chlorobenzophenone 98%
2-Chlorobenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₉ClO
MW: 216.67 g/mol
MDL Number: MFCD00000558
CAS Number: 5162-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR