2,4-Bis(methylthio)pyrimidine 97%

Supplier: Ambeed
Warning

A252363-1G A252363-250MG
77140-508EA 24.73 USD
77140-508 77140-510
2,4-Bis(methylthio)pyrimidine 97%
2,4-Bis(methylthio)pyrimidine

Precaución: for Research Use Only

Formula: C₆H₈N₂S₂
MW: 172.271 g/mol
MDL Number: MFCD18974850
CAS Number: 5909-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR