1,1'-Carbonyldipyrrolidine 98%

Supplier: Ambeed
Warning

Synonyms: Di(pyrrolidin-1-yl)methanone

A251819-25G A251819-5G
77140-494EA 58.35 USD
77140-494 77140-496
1,1'-Carbonyldipyrrolidine 98%
1,1'-Carbonyldipyrrolidine

Precaución: for Research Use Only

Formula: C₉H₁₆N₂O
MW: 168.24 g/mol
MDL Number: MFCD00216641
CAS Number: 81759-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR