(3-Nitro-4-(trifluoromethoxy)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

A251213-100MG A251213-1G A251213-250MG
77140-488EA 37.14 USD
77140-488 77140-490 77140-492
(3-Nitro-4-(trifluoromethoxy)phenyl)boronic acid 97%
(3-Nitro-4-(trifluoromethoxy)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₅BF₃NO₅
MW: 250.93 g/mol
MDL Number: MFCD21332921
CAS Number: 2096333-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR