Dichloro(1,10-phenanthroline)copper(II) 98%

Supplier: Ambeed
Warning

Synonyms: (1,10-Phenanthroline)copper(II) Dichloride, Dichloro(1,10-phenanthroline)copper

A251165-1G A251165-25G A251165-5G
77140-482EA 29.71 USD
77140-482 77140-484 77140-486
Dichloro(1,10-phenanthroline)copper(II) 98%
Dichloro(1,10-phenanthroline)copper(II)

Precaución: for Research Use Only

Formula: C₁₂H₈Cl₂CuN₂
MW: 314.66 g/mol
MDL Number: MFCD00134224
CAS Number: 14783-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR