6-Bromo-2,3-difluorobenzonitrile 97%

Supplier: Ambeed
Warning

A250764-1G A250764-250MG A250764-5G
77140-476EA 27.81 USD
77140-476 77140-478 77140-480
6-Bromo-2,3-difluorobenzonitrile 97%
6-Bromo-2,3-difluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₂BrF₂N
MDL Number: MFCD23381286
CAS Number: 1207875-87-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR