1-(5-Fluoro-6-methoxypyridin-3-yl)ethanone 97%

Supplier: Ambeed
Warning

A249635-100MG A249635-1G A249635-250MG
77140-470EA 31.83 USD
77140-470 77140-472 77140-474
1-(5-Fluoro-6-methoxypyridin-3-yl)ethanone 97%
1-(5-Fluoro-6-methoxypyridin-3-yl)ethanone

Precaución: for Research Use Only

Formula: C₈H₈FNO₂
MW: 169.15 g/mol
MDL Number: MFCD18254435
CAS Number: 1256835-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR