(R)-3-Amino-4-methylpentanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: H-D-β-HoVal-OH

A248796-1G A248796-250MG
77140-458EA 112.46 USD
77140-458 77140-460
(R)-3-Amino-4-methylpentanoic acid 95%
(R)-3-Amino-4-methylpentanoic acid

Precaución: for Research Use Only

Formula: C₆H₁₃NO₂
MW: 131.173 g/mol
MDL Number: MFCD01076231
CAS Number: 75992-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR