5-Bromo-2-methoxynicotinic acid 97%

Supplier: Ambeed
Warning

A247541-100G A247541-10G A247541-1G A247541-250MG A247541-25G A247541-5G
77140-414EA 275.01 USD
77140-414 77140-416 77140-418 77140-420 77140-422 77140-424
5-Bromo-2-methoxynicotinic acid 97%
5-Bromo-2-methoxynicotinic acid

Precaución: for Research Use Only

Formula: C₇H₆BrNO₃
MDL Number: MFCD06656771
CAS Number: 54916-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR