2-Bromo-3-fluoro-6-nitrobenzoic acid 98%

Supplier: Ambeed
Warning

A247529-100MG A247529-1G A247529-250MG
77140-408EA 33.95 USD
77140-408 77140-410 77140-412
2-Bromo-3-fluoro-6-nitrobenzoic acid 98%
2-Bromo-3-fluoro-6-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrFNO₄
MW: 264.01 g/mol
MDL Number: MFCD27931341
CAS Number: 881415-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR