3'-(O-Methyl)adenosine 97%

Supplier: Ambeed
Warning

Synonyms: (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)-4-methoxytetrahydrofuran-3-ol, 3’-O-Methyladenosine

A246201-100MG A246201-1G A246201-250MG
77140-388EA 76.39 USD
77140-388 77140-390 77140-392
3'-(O-Methyl)adenosine 97%
3'-(O-Methyl)adenosine

Precaución: for Research Use Only

Formula: C₁₁H₁₅N₅O₄
MW: 281.2679 g/mol
MDL Number: MFCD00057003
CAS Number: 10300-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR