Mesityleneruthenium(II) chloride Dimer 97%

Supplier: Ambeed
Warning

Synonyms: Dichloro(mesitylene)ruthenium(II) Dimer

A245881-100MG A245881-1G A245881-250MG
77140-382EA 28.84 USD
77140-382 77140-384 77140-386
Mesityleneruthenium(II) chloride Dimer 97%
Mesityleneruthenium(II) chloride Dimer

Precaución: for Research Use Only

Formula: C₁₈H₂₄Cl₄Ru₂
MDL Number: MFCD11656079
CAS Number: 52462-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR