4-Chloro-3-nitroanisole 98%

Supplier: Ambeed
Warning

Synonyms: 1-Chloro-4-methoxy-2-nitrobenzene

A245199-100G A245199-10G A245199-5G
77140-376EA 42.44 USD
77140-376 77140-378 77140-380
4-Chloro-3-nitroanisole 98%
4-Chloro-3-nitroanisole

Precaución: for Research Use Only

Formula: C₇H₆ClNO₃
MW: 187.58 g/mol
MDL Number: MFCD00007077
CAS Number: 10298-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR