(1S,3R)-3-(tert-Butoxycarbonylamino)cyclopentanecarboxylic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(1R,3S)-N-BOC-1-Aminocyclopentane-3-carboxylic acid, Boc-D-AcPAC, (1S,3R)-3-(Boc-amino)cyclopentanecarboxylic Acid, (1S,3R)-3-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid
A244935-10G
A244935-1G
A244935-250MG
A244935-25G
A244935-5G
77140-366EA
213.21
USD
77140-366
77140-368
77140-370
77140-372
77140-374
(1S,3R)-3-(tert-Butoxycarbonylamino)cyclopentanecarboxylic acid 97%
(1S,3R)-3-(tert-Butoxycarbonylamino)cyclopentanecarboxylic acid
Precaución: for Research Use Only
Formula:
C₁₁H₁₉NO₄ MW: 229.28 g/mol |
MDL Number:
MFCD01320857 CAS Number: 261165-05-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...