2-Chloro-6-((4-methoxybenzyl)oxy)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 97%

Supplier: Ambeed
Warning

A244913-100MG A244913-1G A244913-250MG
77140-360EA 37.14 USD
77140-360 77140-362 77140-364
2-Chloro-6-((4-methoxybenzyl)oxy)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 97%
2-Chloro-6-((4-methoxybenzyl)oxy)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₉H₂₃BClNO₄
MW: 375.65 g/mol
MDL Number: MFCD20441902
CAS Number: 2096331-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR