4-(Trifluoromethyl)piperidine 97%

Supplier: Ambeed
Danger

A244473-1G A244473-250MG A244473-25G A244473-5G
77140-350EA 23.68 USD
77140-350 77140-352 77140-354 77140-356
4-(Trifluoromethyl)piperidine 97%
4-(Trifluoromethyl)piperidine

Precaución: for Research Use Only

Formula: C₆H₁₀F₃N
MDL Number: MFCD00102068
CAS Number: 657-36-3
UN: 2734
ADR: 8,3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR