2-Cyclopenten-1-one 97%

Supplier: Ambeed
Danger

Synonyms: 2-Cyclopentenone, Cyclopenten-2-one, Cyclopent-2-enone

A244433-25G A244433-10G A244433-5G A244433-1G
77140-348EA 71.09 USD
77140-348 77550-564 77568-378 77616-008
2-Cyclopenten-1-one 97%
2-Cyclopenten-1-one

Precaución: For research use only.

Formula: C₅H₆O
MW: 82.1 g/mol
MDL Number: MFCD00001401
CAS Number: 930-30-3
UN: 1224
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR