cis-Dichlorobis(triphenylphosphite)platinum(II) 98%

Supplier: Ambeed
Warning

Synonyms: cis-Bis(triphenylphosphine)palladium (II) chloride, cis-Bis(triphenylphosphine)platinum(II) chloride, Pt(PPh₃)₂Cl₂

A244387-1G A244387-250MG
77140-344EA 124.63 USD
77140-344 77140-346
cis-Dichlorobis(triphenylphosphite)platinum(II) 98%
cis-Dichlorobis(triphenylphosphite)platinum(II)

Precaución: for Research Use Only

Formula: [(C₆H₅)₃P]₂PtCl₂
MW: 701.91
MDL Number: MFCD00010825
CAS Number: 15604-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR