5,5-Dimethyl-3-(4-nitro-3-(trifluoromethyl)phenyl)imidazolidine-2,4-dione 99%

Supplier: Ambeed
Danger

Synonyms: Nilutamide

A242214-10MG A242214-1MG A242214-250MG A242214-25MG A242214-50MG
77140-310EA 6.37 USD
77140-310 77140-312 77140-314 77140-316 77140-318
5,5-Dimethyl-3-(4-nitro-3-(trifluoromethyl)phenyl)imidazolidine-2,4-dione 99%
5,5-Dimethyl-3-(4-nitro-3-(trifluoromethyl)phenyl)imidazolidine-2,4-dione

Precaución: for Research Use Only

Formula: C₁₂H₁₀F₃N₃O₄
MW: 317.22 g/mol
MDL Number: MFCD00864670
CAS Number: 63612-50-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR