4-Methyl-3-nitrobenzenesulfonyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 4-Methyl-3-nitrobenzene-1-sulphonyl chloride

A241972-1G A241972-25G A241972-5G
77140-288EA 28.65 USD
77140-288 77140-290 77140-292
4-Methyl-3-nitrobenzenesulfonyl chloride 98%
4-Methyl-3-nitrobenzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₇H₆ClNO₄S
MW: 235.65 g/mol
MDL Number: MFCD00129811
CAS Number: 616-83-1
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR