6-Bromo-4-fluoro-1H-indazol-3-amine 97%

Supplier: Ambeed
Warning

A241599-1G A241599-5G
77140-284EA 161.26 USD
77140-284 77140-286
6-Bromo-4-fluoro-1H-indazol-3-amine 97%
6-Bromo-4-fluoro-1H-indazol-3-amine

Precaución: for Research Use Only

Formula: C₇H₅BrFN₃
MW: 230.037 g/mol
MDL Number: MFCD24368896
CAS Number: 1227912-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR