Carprofen 98%

Supplier: Ambeed
Danger

Synonyms: 2-(6-Chloro-9H-carbazol-2-yl)propanoic acid, 6-Chloro-alpha-methyl-9H-carbazole-2-acetic Acid

A240973-100G A240973-10G A240973-25G A240973-5G A240973-1G
77140-256EA 584.01 USD
77140-256 77140-258 77140-260 77140-262 77593-540
Carprofen 98%
Carprofen

Precaución: for Research Use Only

Formula: C₁₅H₁₂ClNO₂
MW: 273.72 g/mol
MDL Number: MFCD00079028
CAS Number: 53716-49-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR