(R)-3-((tert-Butoxycarbonyl)amino)-4-(2,4,5-trifluorophenyl)butanoic acid 98%

Supplier: Ambeed
Warning

A240774-100G A240774-1G A240774-25G A240774-500G A240774-5G A240774-10G
77140-246EA 114.33 USD
77140-246 77140-248 77140-250 77140-252 77140-254 77685-671
(R)-3-((tert-Butoxycarbonyl)amino)-4-(2,4,5-trifluorophenyl)butanoic acid 98%
(R)-3-((tert-Butoxycarbonyl)amino)-4-(2,4,5-trifluorophenyl)butanoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₈F₃NO₄
MDL Number: MFCD06659147
CAS Number: 486460-00-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR