5-Methoxy-4-methyl-2-nitrobenzoic acid 97%
Supplier: Ambeed
|
Warning
|
A240726-10G
A240726-5G
77140-242EA
591.99
USD
77140-242
77140-244
5-Methoxy-4-methyl-2-nitrobenzoic acid 97%
5-Methoxy-4-methyl-2-nitrobenzoic acid
Precaución: for Research Use Only
Formula:
C₉H₉NO₅ MW: 211.17 g/mol |
MDL Number:
MFCD28291987 CAS Number: 857599-32-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...