7-Bromo-6-chloroisoquinoline 97%

Supplier: Ambeed
Warning

A240671-100MG A240671-250MG A240671-1G
77140-238EA 21.63 USD
77140-238 77140-240 77568-332
7-Bromo-6-chloroisoquinoline 97%
7-Bromo-6-chloroisoquinoline

Precaución: For research use only.

Formula: C₉H₅BrClN
MDL Number: MFCD20486970
CAS Number: 1307316-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR