(1S,2S)-2-Aminocyclopentanecarboxylic acid hydrochloride 97%

Supplier: Ambeed
Warning

A239549-100MG A239549-250MG
77140-216EA 80.34 USD
77140-216 77140-218
(1S,2S)-2-Aminocyclopentanecarboxylic acid hydrochloride 97%
(1S,2S)-2-Aminocyclopentanecarboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₁NO₂·ClH
MDL Number: MFCD09749902
CAS Number: 359849-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR