2-(4-Methoxyphenoxy)propanoic acid 95%

Supplier: Ambeed
Warning

A238561-1G A238561-5G
77140-212EA 26.79 USD
77140-212 77140-214
2-(4-Methoxyphenoxy)propanoic acid 95%
2-(4-Methoxyphenoxy)propanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₄
MDL Number: MFCD01310545
CAS Number: 13794-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR