(2S,3S,4R,5R)-3,4-Dihydroxy-5-(6-((3-iodobenzyl)amino)-9H-purin-9-yl)-N-methyltetrahydrofuran-2-carboxamide 98%

Supplier: Ambeed
Warning

A238381-10MG A238381-1MG A238381-25MG A238381-2MG A238381-50MG A238381-5MG
77140-200EA 57.29 USD
77140-200 77140-202 77140-204 77140-206 77140-208 77140-210
(2S,3S,4R,5R)-3,4-Dihydroxy-5-(6-((3-iodobenzyl)amino)-9H-purin-9-yl)-N-methyltetrahydrofuran-2-carboxamide 98%
(2S,3S,4R,5R)-3,4-Dihydroxy-5-(6-((3-iodobenzyl)amino)-9H-purin-9-yl)-N-methyltetrahydrofuran-2-carboxamide

Precaución: for Research Use Only

Formula: C₁₈H₁₉IN₆O₄
MW: 510.28 g/mol
MDL Number: MFCD00671785
CAS Number: 152918-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR