tert-Butyl (3R,4R)-3-amino-4-hydroxypyrrolidine-1-carboxylate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Boc-(3S,4S)-3-amino-4-hydroxypyrrolidine
A237019-1G
A237019-250MG
A237019-5G
77140-178EA
136.86
USD
77140-178
77140-180
77140-182
tert-Butyl (3R,4R)-3-amino-4-hydroxypyrrolidine-1-carboxylate 95%
tert-Butyl (3R,4R)-3-amino-4-hydroxypyrrolidine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₉H₁₈N₂O₃ MW: 202.251 g/mol |
MDL Number:
MFCD11111251 CAS Number: 190792-74-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...