2-(Di(adamantan-1-yl)phosphino)-N,N-dimethylaniline 98%

Supplier: Ambeed
Warning

A235944-100MG A235944-1G A235944-250MG A235944-5G
77140-158EA 58.35 USD
77140-158 77140-160 77140-162 77140-164
2-(Di(adamantan-1-yl)phosphino)-N,N-dimethylaniline 98%
2-(Di(adamantan-1-yl)phosphino)-N,N-dimethylaniline

Precaución: for Research Use Only

Formula: C₂₈H₄₀NP
MW: 421.59 g/mol
MDL Number: MFCD17018758
CAS Number: 1219080-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR