(1S,2S)-(+)-1,2-Cyclohexanedicarboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: (1S,2S)-Cyclohexane-1,2-dicarboxylic acid

A235696-25G A235696-100G A235696-5G
77140-156EA 45.62 USD
77140-156 77685-654 77685-655
(1S,2S)-(+)-1,2-Cyclohexanedicarboxylic acid 95%
(1S,2S)-(+)-1,2-Cyclohexanedicarboxylic acid

Precaución: for Research Use Only

Formula: C₈H₁₂O₄
MW: 172.18 g/mol
MDL Number: MFCD00151258
CAS Number: 21963-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR