Ethyl 4-phenylthiazole-5-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A234768-100MG
A234768-1G
A234768-250MG
77140-140EA
78.51
USD
77140-140
77140-142
77140-144
Ethyl 4-phenylthiazole-5-carboxylate 97%
Ethyl 4-phenylthiazole-5-carboxylate
Precaución: for Research Use Only
Formula:
C₁₂H₁₁NO₂S MW: 233.286 g/mol |
MDL Number:
MFCD12400736 CAS Number: 99822-80-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...