(2,6-Difluoro-4-isopropoxyphenyl)boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2,6-Difluoro-4-isopropyloxyphenylboronic acid

A233815-100MG A233815-1G A233815-250MG
77140-122EA 33.95 USD
77140-122 77140-124 77140-126
(2,6-Difluoro-4-isopropoxyphenyl)boronic acid 97%
(2,6-Difluoro-4-isopropoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BF₂O₃
MW: 215.99 g/mol
MDL Number: MFCD22689791
CAS Number: 2096337-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR