2-(5-Oxo-6-oxa-4-azaspiro[2.4]heptan-4-yl)acetic acid 97%

Supplier: Ambeed
Warning

A233517-100MG A233517-250MG
77140-118EA 28.84 USD
77140-118 77140-120
2-(5-Oxo-6-oxa-4-azaspiro[2.4]heptan-4-yl)acetic acid 97%
2-(5-Oxo-6-oxa-4-azaspiro[2.4]heptan-4-yl)acetic acid

Precaución: for Research Use Only

Formula: C₇H₉NO₄
MDL Number: MFCD29059170
CAS Number: 1956369-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR