5-Fluoro-7-methoxy-4(3H)-quinazolinone 97%

Supplier: Ambeed
Warning

Synonyms: 5-Fluoro-7-methoxyquinazolin-4(3H)-one

A232697-100MG A232697-250MG
77140-108EA 18.04 USD
77140-108 77140-110
5-Fluoro-7-methoxy-4(3H)-quinazolinone 97%
5-Fluoro-7-methoxy-4(3H)-quinazolinone

Precaución: for Research Use Only

Formula: C₉H₇FN₂O₂
MW: 194.162 g/mol
MDL Number: MFCD13191851
CAS Number: 944742-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR