2-Hydroxy-1-morpholinoethanone 97%
Supplier: Ambeed
|
Warning
|
A231662-10G
A231662-1G
A231662-250MG
A231662-5G
A231662-100G
A231662-25G
77140-090EA
16.47
USD
77140-090
77140-092
77140-094
77140-096
77593-502
77685-636
2-Hydroxy-1-morpholinoethanone 97%
2-Hydroxy-1-morpholinoethanone
Precaución: for Research Use Only
Formula:
C₆H₁₁NO₃ MW: 145.16 g/mol |
MDL Number:
MFCD08436413 CAS Number: 51068-78-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...