(3-Chloro-5-fluoro-4-isopropoxyphenyl)boronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-3-fluoro-4-isopropoxyphenylboronic acid

A231603-1G A231603-250MG A231603-5G
77140-084EA 145.35 USD
77140-084 77140-086 77140-088
(3-Chloro-5-fluoro-4-isopropoxyphenyl)boronic acid 95%
(3-Chloro-5-fluoro-4-isopropoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BClFO₃
MW: 232.44 g/mol
MDL Number: MFCD20441800
CAS Number: 2096335-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR