1-((6-Bromopyridin-3-yl)methyl)-4-ethylpiperazine 97%

Supplier: Ambeed
Warning

A231549-10G A231549-1G A231549-5G
77140-078EA 209.09 USD
77140-078 77140-080 77140-082
1-((6-Bromopyridin-3-yl)methyl)-4-ethylpiperazine 97%
1-((6-Bromopyridin-3-yl)methyl)-4-ethylpiperazine

Precaución: for Research Use Only

Formula: C₁₂H₁₈BrN₃
MDL Number: MFCD25977325
CAS Number: 1231930-25-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR