(R)-3-(1-(Dimethylamino)ethyl)phenol 95%
Supplier: Ambeed
|
Warning
|
A231532-10G
A231532-1G
A231532-250MG
A231532-25G
A231532-5G
77140-068EA
248.26
USD
77140-068
77140-070
77140-072
77140-074
77140-076
(R)-3-(1-(Dimethylamino)ethyl)phenol 95%
(R)-3-(1-(Dimethylamino)ethyl)phenol
Precaución: for Research Use Only
Formula:
C₁₀H₁₅NO MW: 165.232 g/mol |
MDL Number:
MFCD06656490 CAS Number: 851086-95-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...